Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
Chromosome Location
Chr.2: 159315312 - 159712439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289975.1 NP_001276904.1 19 - 20 3373 123
NM_013450.3 NP_038478.2 20 - 21 3481 123
XM_005246488.2 XP_005246545.2 21 - 22 3357 123
XM_005246489.4 XP_005246546.2 20 - 21 4460 123
XM_005246492.4 XP_005246549.2 20 - 21 4455 123
XM_006712461.3 XP_006712524.2 24 - 25 3931 123
XM_011511038.2 XP_011509340.1 21 - 22 3455 123
XM_011511039.1 XP_011509341.1 18 - 19 3078 123
XM_011511042.1 XP_011509344.1 19 - 20 3111 123
XM_011511043.1 XP_011509345.1 17 - 18 2976 123
XM_011511044.1 XP_011509346.1 18 - 19 3003 123
XM_011511045.1 XP_011509347.1 18 - 19 2916 123
XM_011511047.1 XP_011509349.1 20 - 21 3263 123
XM_011511048.2 XP_011509350.1 20 - 21 3328 123
XM_011511050.1 XP_011509352.1 18 - 19 2811 123
XM_011511051.1 XP_011509353.1 18 - 19 2997 123
XM_011511052.1 XP_011509354.1 17 - 18 2715 123
XM_011511053.2 XP_011509355.1 19 - 20 3111 123
XM_011511054.2 XP_011509356.1 19 - 20 3111 123
XM_011511056.1 XP_011509358.1 9 - 10 731 123
XM_017003918.1 XP_016859407.1 24 - 25 3921 123
XM_017003919.1 XP_016859408.1 24 - 25 3915 123
XM_017003920.1 XP_016859409.1 26 - 27 4132 123
XM_017003921.1 XP_016859410.1 21 - 22 3455 123
XM_017003922.1 XP_016859411.1 21 - 22 3351 123
XM_017003923.1 XP_016859412.1 19 - 20 3105 123
XM_017003924.1 XP_016859413.1 18 - 19 3003 123
XM_017003925.1 XP_016859414.1 20 - 21 3359 123
XM_017003926.1 XP_016859415.1 20 - 21 3217 123
XM_017003927.1 XP_016859416.1 20 - 21 3257 123
XM_017003928.1 XP_016859417.1 20 - 21 3167 123
XM_017003929.1 XP_016859418.1 20 - 21 3249 123
XM_017003930.1 XP_016859419.1 20 - 21 3057 123
XM_017003931.1 XP_016859420.1 20 - 21 3155 123
XM_017003932.1 XP_016859421.1 17 - 18 2808 123
XM_017003933.1 XP_016859422.1 19 - 20 3034 123
XM_017003934.1 XP_016859423.1 19 - 20 2929 123
XM_017003935.1 XP_016859424.1 18 - 19 2867 123
XM_017003936.1 XP_016859425.1 18 - 19 2821 123
XM_017003937.1 XP_016859426.1 13 - 14 1528 123
GenBank mRNA AB032255.1 - 15 - 16 2854 123
AB040909.2 - 19 - 20 3341 123
AK027612.1 - 11 - 12 1108 123

View Full Product Details