Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
Chromosome Location
Chr.2: 159315312 - 159712439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289975.1 NP_001276904.1 30 - 31 5749 140
NM_013450.3 NP_038478.2 31 - 32 5857 140
XM_005246488.2 XP_005246545.2 32 - 33 5733 140
XM_005246489.4 XP_005246546.2 31 - 32 6836 140
XM_005246492.4 XP_005246549.2 31 - 32 6831 140
XM_006712461.3 XP_006712524.2 35 - 36 6307 140
XM_011511038.2 XP_011509340.1 32 - 33 5831 140
XM_011511039.1 XP_011509341.1 29 - 30 5454 140
XM_011511042.1 XP_011509344.1 30 - 31 5487 140
XM_011511043.1 XP_011509345.1 28 - 29 5352 140
XM_011511044.1 XP_011509346.1 29 - 30 5379 140
XM_011511045.1 XP_011509347.1 29 - 30 5292 140
XM_011511047.1 XP_011509349.1 31 - 32 5639 140
XM_011511048.2 XP_011509350.1 31 - 32 5704 140
XM_011511050.1 XP_011509352.1 29 - 30 5187 140
XM_011511051.1 XP_011509353.1 29 - 30 5373 140
XM_011511052.1 XP_011509354.1 28 - 29 5091 140
XM_011511053.2 XP_011509355.1 30 - 31 5487 140
XM_011511054.2 XP_011509356.1 30 - 31 5487 140
XM_011511056.1 XP_011509358.1 20 - 21 3107 140
XM_017003918.1 XP_016859407.1 35 - 36 6297 140
XM_017003919.1 XP_016859408.1 35 - 36 6291 140
XM_017003920.1 XP_016859409.1 37 - 38 6508 140
XM_017003921.1 XP_016859410.1 32 - 33 5831 140
XM_017003922.1 XP_016859411.1 32 - 33 5727 140
XM_017003923.1 XP_016859412.1 30 - 31 5481 140
XM_017003924.1 XP_016859413.1 29 - 30 5379 140
XM_017003925.1 XP_016859414.1 31 - 32 5735 140
XM_017003926.1 XP_016859415.1 31 - 32 5593 140
XM_017003927.1 XP_016859416.1 31 - 32 5633 140
XM_017003928.1 XP_016859417.1 31 - 32 5543 140
XM_017003929.1 XP_016859418.1 31 - 32 5625 140
XM_017003930.1 XP_016859419.1 31 - 32 5433 140
XM_017003931.1 XP_016859420.1 31 - 32 5531 140
XM_017003932.1 XP_016859421.1 28 - 29 5184 140
XM_017003933.1 XP_016859422.1 30 - 31 5410 140
XM_017003934.1 XP_016859423.1 30 - 31 5305 140
XM_017003935.1 XP_016859424.1 29 - 30 5243 140
XM_017003936.1 XP_016859425.1 29 - 30 5197 140
XM_017003937.1 XP_016859426.1 24 - 25 3904 140
GenBank mRNA AB032255.1 - 26 - 27 5230 140
AB040909.2 - 30 - 31 5717 140
AK074895.1 - 9 - 10 1975 140
AL080173.1 - 2 - 3 297 140

View Full Product Details