The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142279.2 | NP_001135751.1 | 1 - 2 | 458 | 113 |
NM_024570.3 | NP_078846.2 | 1 - 2 | 458 | 113 | |
XM_011535229.1 | XP_011533531.1 | 1 - 2 | 380 | 113 | |
XM_011535230.2 | XP_011533532.1 | 1 - 2 | 380 | 113 | |
XM_011535231.2 | XP_011533533.1 | 1 - 2 | 380 | 113 | |
XM_011535233.2 | XP_011533535.1 | 1 - 2 | 128 | 113 | |
XM_017020747.1 | XP_016876236.1 | 1 - 2 | 380 | 113 | |
GenBank mRNA | AK021774.1 | - | 1 - 2 | 345 | 113 |
AK223340.1 | - | 1 - 2 | 345 | 113 | |
AY764036.1 | - | 1 - 2 | 366 | 113 | |
BC001397.2 | - | 1 - 2 | 168 | 113 | |
BC005088.1 | - | 1 - 2 | 168 | 113 | |
BC007332.2 | - | 1 - 2 | 168 | 113 | |
BC010174.2 | - | 1 - 2 | 168 | 113 | |
BC036744.1 | - | 1 - 2 | 329 | 113 | |
BX382357.2 | - | 1 - 2 | 336 | 113 | |
DB221997.1 | - | 1 - 2 | 380 | 113 | |
DT216141.1 | - | 1 - 2 | 380 | 113 |