The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142279.2 | NP_001135751.1 | 5 - 6 | 842 | 147 |
NM_024570.3 | NP_078846.2 | 5 - 6 | 842 | 147 | |
XM_006719867.3 | XP_006719930.1 | 5 - 6 | 531 | 147 | |
XM_011535229.1 | XP_011533531.1 | 5 - 6 | 764 | 147 | |
XM_011535230.2 | XP_011533532.1 | 5 - 6 | 764 | 147 | |
XM_011535231.2 | XP_011533533.1 | 5 - 6 | 764 | 147 | |
XM_017020747.1 | XP_016876236.1 | 5 - 6 | 764 | 147 | |
GenBank mRNA | AK021774.1 | - | 5 - 6 | 729 | 147 |
AK223340.1 | - | 5 - 6 | 729 | 147 | |
AY764036.1 | - | 5 - 6 | 750 | 147 | |
BC001397.2 | - | 5 - 6 | 552 | 147 | |
BC005088.1 | - | 5 - 6 | 552 | 147 | |
BC007332.2 | - | 5 - 6 | 552 | 147 | |
BC010174.2 | - | 5 - 6 | 552 | 147 | |
BC036744.1 | - | 5 - 6 | 713 | 147 | |
BX328027.1 | - | 2 - 3 | 169 | 147 |