Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM1A
Entrez Gene ID
23028
Gene Name
lysine demethylase 1A
Gene Aliases
AOF2, BHC110, CPRF, KDM1, LSD1
Chromosome Location
Chr.1: 23019443 - 23083691 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009999.2 NP_001009999.1 19 - 20 2449 79
NM_015013.3 NP_055828.2 17 - 18 2377 79
XM_005245786.2 XP_005245843.1 18 - 19 2442 79
XM_006710472.3 XP_006710535.1 17 - 18 2400 79
XM_006710473.2 XP_006710536.1 18 - 19 2460 79
XM_006710474.3 XP_006710537.1 18 - 19 2460 79
XM_017000716.1 XP_016856205.1 18 - 19 2442 79
XM_017000717.1 XP_016856206.1 17 - 18 2382 79
XM_017000718.1 XP_016856207.1 16 - 17 1866 79
XM_017000719.1 XP_016856208.1 17 - 18 1992 79
XM_017000720.1 XP_016856209.1 16 - 17 1962 79
GenBank mRNA AB011173.1 - 17 - 18 2330 79
AK290328.1 - 17 - 18 1903 79
AL833812.1 - 13 - 14 1499 79
BC016639.1 - 15 - 16 1616 79
BC025362.1 - 9 - 10 1042 79
BC040194.2 - 19 - 20 2404 79
BC048134.1 - 17 - 18 2298 79

View Full Product Details