Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM1A
Entrez Gene ID
23028
Gene Name
lysine demethylase 1A
Gene Aliases
AOF2, BHC110, CPRF, KDM1, LSD1
Chromosome Location
Chr.1: 23019443 - 23083691 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009999.2 NP_001009999.1 4 - 5 862 96
NM_015013.3 NP_055828.2 3 - 4 802 96
XM_005245786.2 XP_005245843.1 4 - 5 867 96
XM_006710472.3 XP_006710535.1 3 - 4 807 96
XM_006710473.2 XP_006710536.1 4 - 5 867 96
XM_006710474.3 XP_006710537.1 4 - 5 867 96
XM_017000716.1 XP_016856205.1 4 - 5 867 96
XM_017000717.1 XP_016856206.1 3 - 4 807 96
GenBank mRNA AB011173.1 - 3 - 4 755 96
AK290328.1 - 3 - 4 328 96
BC040194.2 - 4 - 5 817 96
BC048134.1 - 3 - 4 723 96

View Full Product Details