The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310121.1 | NP_001297050.1 | 13 - 14 | 1710 | 136 |
NM_001310123.1 | NP_001297052.1 | 11 - 12 | 1601 | 136 | |
NM_002973.3 | NP_002964.3 | 13 - 14 | 2504 | 136 | |
NR_132311.1 | 13 - 14 | 2504 | 136 | ||
GenBank mRNA | BC111757.1 | - | 13 - 14 | 1710 | 136 |
BC114546.1 | - | 11 - 12 | 1601 | 136 | |
U70323.1 | - | 13 - 14 | 2501 | 136 | |
Y08262.1 | - | 13 - 14 | 2101 | 136 |