The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310121.1 | NP_001297050.1 | 21 - 22 | 2731 | 97 |
NM_002973.3 | NP_002964.3 | 21 - 22 | 3525 | 97 | |
NR_132311.1 | 20 - 21 | 3464 | 97 | ||
GenBank mRNA | BC111757.1 | - | 21 - 22 | 2731 | 97 |
U70323.1 | - | 21 - 22 | 3522 | 97 | |
Y08262.1 | - | 20 - 21 | 3061 | 97 |