Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATXN2
Entrez Gene ID
6311
Gene Name
ataxin 2
Gene Aliases
ASL13, ATX2, SCA2, TNRC13
Chromosome Location
Chr.12: 111452214 - 111599676 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001310121.1 NP_001297050.1 3 - 4 197 157
NM_002973.3 NP_002964.3 3 - 4 991 157
NR_132311.1 3 - 4 991 157
GenBank mRNA AK307803.1 - 3 - 4 423 157
BC111757.1 - 3 - 4 197 157
U70323.1 - 3 - 4 988 157
Y08262.1 - 3 - 4 588 157

View Full Product Details