Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEMT
Entrez Gene ID
10400
Gene Name
phosphatidylethanolamine N-methyltransferase
Gene Aliases
PEAMT, PEMPT, PEMT2, PNMT
Chromosome Location
Chr.17: 17505561 - 17591703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267551.1 NP_001254480.1 6 - 7 669 122
NM_007169.2 NP_009100.2 6 - 7 727 122
NM_148172.2 NP_680477.1 6 - 7 735 122
NM_148173.1 NP_680478.1 6 - 7 614 122
XM_006721418.3 XP_006721481.2 6 - 7 616 122
XM_017024016.1 XP_016879505.1 4 - 5 522 122
GenBank mRNA AB029821.1 - 6 - 7 727 122
AF044214.1 - 6 - 7 700 122
AF113126.1 - 6 - 7 704 122
AF176806.1 - 6 - 7 677 122
AF176807.1 - 6 - 7 622 122
BC000557.2 - 6 - 7 711 122
BC050593.1 - 6 - 7 712 122
BM011377.1 - 6 - 7 597 122
CR457099.1 - 5 - 6 547 122

View Full Product Details