Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACOT12
Entrez Gene ID
134526
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
CACH-1, Cach, STARD15, THEAL
Chromosome Location
Chr.5: 81309045 - 81394174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130767.2 NP_570123.1 5 - 6 550 108
XM_006714531.2 XP_006714594.1 5 - 6 515 108
XM_006714532.3 XP_006714595.1 5 - 6 515 108
XM_017009045.1 XP_016864534.1 5 - 6 515 108
XM_017009046.1 XP_016864535.1 5 - 6 515 108
XM_017009047.1 XP_016864536.1 4 - 5 453 108
GenBank mRNA AB078619.1 - 5 - 6 518 108
AK122960.1 - 5 - 6 515 108
BC075010.2 - 5 - 6 516 108
BC075011.2 - 5 - 6 511 108

View Full Product Details