Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
Chromosome Location
Chr.2: 233251571 - 233295674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190266.1 NP_001177195.1 7 - 8 1047 53
NM_001190267.1 NP_001177196.1 7 - 8 1045 53
NM_030803.6 NP_110430.5 7 - 8 1049 53
XM_005246082.2 XP_005246139.1 7 - 8 986 53
XM_005246084.2 XP_005246141.1 4 - 5 525 53
XM_005246086.2 XP_005246143.1 4 - 5 525 53
XM_006712608.2 XP_006712671.1 6 - 7 797 53
GenBank mRNA AK000897.1 - 6 - 7 660 53
AK123876.1 - 4 - 5 516 53
AY358182.1 - 7 - 8 938 53
AY398617.1 - 7 - 8 938 53
BC117337.1 - 7 - 8 711 53

View Full Product Details