The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005783.2 | NP_001005783.2 | 3 - 4 | 490 | 73 |
NM_016527.3 | NP_057611.1 | 2 - 3 | 270 | 73 | |
XM_011541558.1 | XP_011539860.1 | 1 - 2 | 143 | 73 | |
XM_011541559.1 | XP_011539861.1 | 1 - 2 | 143 | 73 | |
XM_011541560.1 | XP_011539862.1 | 1 - 2 | 143 | 73 | |
XM_011541561.1 | XP_011539863.1 | 1 - 2 | 143 | 73 | |
XM_017001421.1 | XP_016856910.1 | 3 - 4 | 826 | 73 | |
GenBank mRNA | AF203975.1 | - | 3 - 4 | 397 | 73 |
AF231917.1 | - | 2 - 3 | 208 | 73 | |
AK298289.1 | - | 3 - 4 | 490 | 73 | |
BC020863.1 | - | 3 - 4 | 457 | 73 | |
BG428999.1 | - | 2 - 3 | 266 | 73 | |
CB121424.1 | - | 2 - 3 | 267 | 73 |