Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCAM1
Entrez Gene ID
7412
Gene Name
vascular cell adhesion molecule 1
Gene Aliases
CD106, INCAM-100
Chromosome Location
Chr.1: 100719640 - 100739045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078.3 NP_001069.1 3 - 4 878 133
NM_001199834.1 NP_001186763.1 3 - 4 692 133
NM_080682.2 NP_542413.1 3 - 4 878 133
GenBank mRNA AK223266.1 - 3 - 4 777 133
AK291732.1 - 3 - 4 776 133
AK296692.1 - 3 - 4 590 133
BC017276.2 - 3 - 4 758 133
BC068490.1 - 3 - 4 776 133
BC085003.1 - 3 - 4 758 133
M30257.1 - 3 - 4 763 133
M60335.1 - 3 - 4 756 133
X53051.1 - 3 - 4 657 133

View Full Product Details