The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007065.3 | NP_008996.1 | 3 - 4 | 604 | 106 |
XM_011527652.2 | XP_011525954.1 | 3 - 4 | 724 | 106 | |
GenBank mRNA | BC000083.2 | - | 3 - 4 | 528 | 106 |
BC008793.2 | - | 3 - 4 | 528 | 106 | |
BE796008.1 | - | 3 - 4 | 608 | 107 | |
BT006796.1 | - | 3 - 4 | 488 | 106 | |
CR542249.1 | - | 3 - 4 | 488 | 106 | |
U43077.1 | - | 3 - 4 | 496 | 106 | |
U63131.1 | - | 3 - 4 | 536 | 106 |