Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTN4
Entrez Gene ID
59277
Gene Name
netrin 4
Gene Aliases
PRO3091
Chromosome Location
Chr.12: 95657807 - 95790758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021229.3 NP_067052.2 2 - 3 1029 114
GenBank mRNA AF119916.3 - 3 - 4 566 114
AF278532.1 - 2 - 3 1031 114
AF297711.1 - 2 - 3 881 114
AK024691.1 - 1 - 2 105 114
AK226070.1 - 2 - 3 886 114
AK290591.1 - 2 - 3 566 114
AK315796.1 - 2 - 3 888 114
AL833767.1 - 1 - 2 433 114
AY330211.1 - 2 - 3 580 114
BC013591.1 - 1 - 2 104 114
BC136798.1 - 2 - 3 1004 114
BC136799.1 - 2 - 3 1004 114

View Full Product Details