The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145071.2 | NP_659508.1 | 1 - 2 | 198 | 121 |
GenBank mRNA | AK313850.1 | - | 1 - 2 | 141 | 121 |
BC031590.1 | - | 1 - 2 | 130 | 121 | |
D83532.1 | - | 1 - 2 | 128 | 121 | |
DA957793.1 | - | 1 - 2 | 198 | 121 |