The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017515.4 | NP_059985.2 | 2 - 3 | 704 | 128 |
GenBank mRNA | AK023080.1 | - | 2 - 3 | 301 | 128 |
AK128062.1 | - | 5 - 6 | 873 | 128 | |
AL833969.1 | - | 2 - 3 | 307 | 128 | |
BC039195.2 | - | 2 - 3 | 444 | 128 | |
BC048302.1 | - | 2 - 3 | 310 | 128 | |
X99961.1 | - | 2 - 3 | 247 | 128 |