The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017515.4 | NP_059985.2 | 4 - 5 | 996 | 76 |
GenBank mRNA | AK023080.1 | - | 4 - 5 | 593 | 76 |
AK128062.1 | - | 7 - 8 | 1165 | 76 | |
AK300582.1 | - | 3 - 4 | 431 | 76 | |
BC039195.2 | - | 4 - 5 | 736 | 76 | |
BC048302.1 | - | 4 - 5 | 602 | 76 |