Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTUB1
Entrez Gene ID
55611
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
HSPC263, OTB1, OTU1
Chromosome Location
Chr.11: 63985853 - 63998420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017670.2 NP_060140.2 5 - 6 1032 76
NR_003089.1 4 - 5 1240 76
GenBank mRNA AF161381.1 - 5 - 6 440 76
AK098029.1 - 4 - 5 1240 76
AK225133.1 - 5 - 6 439 76
AK303228.1 - 4 - 5 682 76
AK307413.1 - 5 - 6 567 76
AL096714.1 - 4 - 5 651 76
AY177200.1 - 5 - 6 428 76
BC007519.2 - 5 - 6 436 76
BC010368.1 - 5 - 6 458 76
BC107701.1 - 5 - 6 458 76

View Full Product Details