Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC38
Entrez Gene ID
55020
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
LL22NC03-5H6.5
Chromosome Location
Chr.22: 46267964 - 46294008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017931.2 NP_060401.2 11 - 12 1159 125
XM_011530259.1 XP_011528561.1 12 - 13 1287 125
GenBank mRNA AK000706.1 - 11 - 12 1114 125
AK300606.1 - 10 - 11 965 125
BC018918.2 - 11 - 12 1100 125

View Full Product Details