Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPCPD1
Entrez Gene ID
56261
Gene Name
glycerophosphocholine phosphodiesterase 1
Gene Aliases
EDI3, GDE5, GDPD6, PREI4
Chromosome Location
Chr.20: 5544434 - 5611026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019593.3 NP_062539.1 17 - 18 1747 143
XM_005260758.4 XP_005260815.1 18 - 19 1898 143
XM_005260761.1 XP_005260818.1 16 - 17 1671 143
XM_006723596.3 XP_006723659.1 18 - 19 2671 143
GenBank mRNA AB037855.1 - 17 - 18 1738 143
AK001947.1 - 4 - 5 300 143
AL833069.1 - 12 - 13 2943 143
BC027588.2 - 17 - 18 1746 143

View Full Product Details