Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr.13: 45954469 - 46052785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015070.4 NP_055885.3 12 - 13 2538 90
XM_005266301.3 XP_005266358.1 13 - 14 2916 90
XM_005266302.3 XP_005266359.1 13 - 14 2616 90
XM_005266303.3 XP_005266360.1 14 - 15 2941 90
XM_005266304.3 XP_005266361.1 13 - 14 2898 90
XM_005266305.3 XP_005266362.1 14 - 15 2934 90
XM_005266306.3 XP_005266363.1 13 - 14 2612 90
XM_005266307.3 XP_005266364.1 13 - 14 2916 90
XM_005266308.3 XP_005266365.1 12 - 13 2832 90
XM_005266309.3 XP_005266366.1 12 - 13 2532 90
XM_005266310.3 XP_005266367.1 12 - 13 2814 90
XM_005266311.3 XP_005266368.1 12 - 13 2832 90
XM_005266312.1 XP_005266369.1 12 - 13 2832 90
XM_017020475.1 XP_016875964.1 14 - 15 2704 90
XM_017020476.1 XP_016875965.1 14 - 15 2701 90
XM_017020477.1 XP_016875966.1 13 - 14 2857 90
XM_017020478.1 XP_016875967.1 13 - 14 2850 90
XM_017020479.1 XP_016875968.1 13 - 14 2620 90
XM_017020480.1 XP_016875969.1 12 - 13 2528 90
XM_017020481.1 XP_016875970.1 13 - 14 2857 90
XM_017020482.1 XP_016875971.1 12 - 13 2532 90
GenBank mRNA AB020660.1 - 1 - 2 364 90
AK056422.1 - 12 - 13 2813 90
AK092988.1 - 9 - 10 2177 90
AL136745.1 - 5 - 6 1574 90
AL831833.1 - 7 - 8 1942 90
AY283618.1 - 12 - 13 2536 90

View Full Product Details