Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr.13: 45954469 - 46052785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015070.4 NP_055885.3 15 - 16 4388 89
XM_005266301.3 XP_005266358.1 16 - 17 4766 89
XM_005266302.3 XP_005266359.1 16 - 17 4466 89
XM_005266303.3 XP_005266360.1 17 - 18 4791 89
XM_005266304.3 XP_005266361.1 16 - 17 4748 89
XM_005266305.3 XP_005266362.1 17 - 18 4784 89
XM_005266306.3 XP_005266363.1 16 - 17 4462 89
XM_005266308.3 XP_005266365.1 15 - 16 4682 89
XM_005266309.3 XP_005266366.1 15 - 16 4382 89
XM_005266310.3 XP_005266367.1 15 - 16 4664 89
XM_005266312.1 XP_005266369.1 15 - 16 4682 89
XM_017020475.1 XP_016875964.1 17 - 18 4554 89
XM_017020476.1 XP_016875965.1 17 - 18 4551 89
XM_017020477.1 XP_016875966.1 16 - 17 4707 89
XM_017020478.1 XP_016875967.1 16 - 17 4700 89
XM_017020479.1 XP_016875968.1 16 - 17 4470 89
XM_017020480.1 XP_016875969.1 15 - 16 4378 89
GenBank mRNA AK001815.1 - 1 - 2 73 89
AL136745.1 - 8 - 9 3424 89
AY283618.1 - 15 - 16 4386 89
CN262326.1 - 2 - 3 616 89

View Full Product Details