Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr.13: 45954469 - 46052785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015070.4 NP_055885.3 7 - 8 822 82
XM_005266301.3 XP_005266358.1 7 - 8 1116 82
XM_005266302.3 XP_005266359.1 7 - 8 816 82
XM_005266303.3 XP_005266360.1 8 - 9 1141 82
XM_005266304.3 XP_005266361.1 7 - 8 1098 82
XM_005266305.3 XP_005266362.1 8 - 9 1134 82
XM_005266306.3 XP_005266363.1 7 - 8 812 82
XM_005266307.3 XP_005266364.1 7 - 8 1116 82
XM_005266308.3 XP_005266365.1 7 - 8 1116 82
XM_005266309.3 XP_005266366.1 7 - 8 816 82
XM_005266310.3 XP_005266367.1 7 - 8 1098 82
XM_005266311.3 XP_005266368.1 7 - 8 1116 82
XM_005266312.1 XP_005266369.1 7 - 8 1116 82
XM_017020475.1 XP_016875964.1 8 - 9 904 82
XM_017020476.1 XP_016875965.1 8 - 9 901 82
XM_017020477.1 XP_016875966.1 8 - 9 1141 82
XM_017020478.1 XP_016875967.1 8 - 9 1134 82
XM_017020479.1 XP_016875968.1 8 - 9 904 82
XM_017020480.1 XP_016875969.1 7 - 8 812 82
XM_017020481.1 XP_016875970.1 8 - 9 1141 82
XM_017020482.1 XP_016875971.1 7 - 8 816 82
GenBank mRNA AK056422.1 - 7 - 8 1099 82
AK092988.1 - 4 - 5 461 82
AL831833.1 - 2 - 3 226 82
AY283618.1 - 7 - 8 820 82
BC019000.2 - 7 - 8 1063 82
BC050658.2 - 4 - 5 491 82

View Full Product Details