The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271821.1 | NP_001258750.1 | 2 - 3 | 494 | 91 |
NM_018965.3 | NP_061838.1 | 2 - 3 | 494 | 91 | |
GenBank mRNA | AB062787.1 | - | 2 - 3 | 494 | 91 |
AF213457.1 | - | 2 - 3 | 484 | 91 | |
AK312215.1 | - | 2 - 3 | 467 | 91 | |
BC032362.1 | - | 2 - 3 | 411 | 91 |