The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031431.3 | NP_113619.2 | 21 - 22 | 2501 | 138 |
XM_011535266.1 | XP_011533568.1 | 21 - 22 | 2308 | 138 | |
XM_017020790.1 | XP_016876279.1 | 20 - 21 | 2154 | 138 | |
GenBank mRNA | AF111707.1 | - | 1 - 2 | 87 | 138 |
AF332595.1 | - | 21 - 22 | 2460 | 138 | |
AF349676.1 | - | 21 - 22 | 2480 | 138 | |
AK026994.1 | - | 4 - 5 | 434 | 138 | |
AK074374.1 | - | 9 - 10 | 1045 | 138 | |
AK304248.1 | - | 21 - 22 | 2303 | 138 | |
AK314387.1 | - | 21 - 22 | 2427 | 138 | |
BX648211.1 | - | 21 - 22 | 2064 | 138 |