The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031431.3 | NP_113619.2 | 2 - 3 | 464 | 148 |
XM_011535266.1 | XP_011533568.1 | 2 - 3 | 392 | 148 | |
XM_011535267.2 | XP_011533569.1 | 2 - 3 | 444 | 148 | |
XM_017020790.1 | XP_016876279.1 | 1 - 2 | 148 | 148 | |
GenBank mRNA | AF332595.1 | - | 2 - 3 | 423 | 148 |
AK026305.1 | - | 2 - 3 | 423 | 148 | |
AK225466.1 | - | 2 - 3 | 423 | 148 | |
AK225952.1 | - | 2 - 3 | 423 | 148 | |
AK294959.1 | - | 2 - 3 | 393 | 148 | |
AK304248.1 | - | 2 - 3 | 391 | 148 | |
AK314387.1 | - | 2 - 3 | 390 | 148 | |
BC038953.1 | - | 2 - 3 | 354 | 148 | |
BX648211.1 | - | 2 - 3 | 148 | 148 | |
DC307455.1 | - | 2 - 3 | 466 | 148 |