The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165962.1 | NP_001159434.1 | 10 - 11 | 982 | 78 |
NM_018127.6 | NP_060597.4 | 11 - 12 | 1102 | 78 | |
NM_173717.1 | NP_776065.1 | 11 - 12 | 1102 | 78 | |
GenBank mRNA | AF304370.1 | - | 11 - 12 | 985 | 78 |
AK001392.1 | - | 11 - 12 | 1059 | 78 | |
AK074244.1 | - | 3 - 4 | 224 | 78 | |
AK094333.1 | - | 3 - 4 | 813 | 78 | |
AK094687.1 | - | 11 - 12 | 986 | 78 | |
AK125030.1 | - | 10 - 11 | 1247 | 78 | |
AK298397.1 | - | 10 - 11 | 917 | 78 | |
AK300011.1 | - | 11 - 12 | 977 | 78 | |
BC001939.1 | - | 11 - 12 | 1045 | 78 | |
BC004158.2 | - | 11 - 12 | 1045 | 78 | |
CR457261.1 | - | 11 - 12 | 985 | 78 | |
KU178737.1 | - | 11 - 12 | 985 | 78 |