Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELAC2
Entrez Gene ID
60528
Gene Name
elaC ribonuclease Z 2
Gene Aliases
COXPD17, ELC2, HPC2
Chromosome Location
Chr.17: 12991612 - 13018064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165962.1 NP_001159434.1 20 - 21 2025 101
NM_018127.6 NP_060597.4 21 - 22 2145 101
NM_173717.1 NP_776065.1 21 - 22 2142 101
GenBank mRNA AF304370.1 - 21 - 22 2028 101
AK001392.1 - 21 - 22 2099 101
AK074244.1 - 13 - 14 1267 101
AK094012.1 - 6 - 7 1915 101
AK094333.1 - 13 - 14 1856 101
AK094687.1 - 21 - 22 2029 101
AK124838.1 - 10 - 11 1574 101
AK125030.1 - 19 - 20 2858 101
AK298397.1 - 20 - 21 1960 101
AK300011.1 - 21 - 22 2020 101
AK304589.1 - 16 - 17 1427 101
BC001939.1 - 21 - 22 2088 101
BC004158.2 - 21 - 22 2088 101
CR457261.1 - 21 - 22 2028 101
KU178737.1 - 21 - 22 2028 101

View Full Product Details