The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001420.3 | NP_001411.2 | 2 - 3 | 607 | 128 |
NM_032281.2 | NP_115657.2 | 2 - 3 | 607 | 128 | |
XM_011527778.2 | XP_011526080.1 | 2 - 3 | 610 | 128 | |
GenBank mRNA | AY034002.1 | - | 1 - 2 | 196 | 128 |
BC014144.2 | - | 2 - 3 | 600 | 128 | |
D26158.1 | - | 2 - 3 | 473 | 128 |