Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KANK1
Entrez Gene ID
23189
Gene Name
KN motif and ankyrin repeat domains 1
Gene Aliases
ANKRD15, CPSQ2, KANK
Chromosome Location
Chr.9: 470294 - 746106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256876.1 NP_001243805.1 15 - 16 4647 75
NM_001256877.1 NP_001243806.1 12 - 13 4756 75
NM_015158.3 NP_055973.2 11 - 12 4141 75
NM_153186.4 NP_694856.1 10 - 11 4403 75
XM_017014514.1 XP_016870003.1 11 - 12 4553 75
XM_017014515.1 XP_016870004.1 12 - 13 4289 75
XM_017014517.1 XP_016870006.1 11 - 12 4499 75
XM_017014518.1 XP_016870007.1 11 - 12 4087 75
XM_017014521.1 XP_016870010.1 10 - 11 4313 75
XM_017014522.1 XP_016870011.1 11 - 12 4114 75
XM_017014523.1 XP_016870012.1 10 - 11 3901 75
XM_017014525.1 XP_016870014.1 10 - 11 4259 75
XM_017014527.1 XP_016870016.1 10 - 11 4333 75
XM_017014528.1 XP_016870017.1 11 - 12 4187 75
XM_017014529.1 XP_016870018.1 10 - 11 4600 75
XM_017014530.1 XP_016870019.1 10 - 11 4109 75
XM_017014534.1 XP_016870023.1 10 - 11 4279 75
XM_017014535.1 XP_016870024.1 10 - 11 4545 75
XM_017014536.1 XP_016870025.1 10 - 11 4248 75
XM_017014539.1 XP_016870028.1 9 - 10 3882 75
XM_017014540.1 XP_016870029.1 9 - 10 4358 75
XM_017014541.1 XP_016870030.1 9 - 10 4362 75
XM_017014542.1 XP_016870031.1 9 - 10 3815 75
GenBank mRNA AK025428.1 - 9 - 10 2286 75
AK290028.1 - 11 - 12 4107 75
AK292989.1 - 11 - 12 4120 75
AL833015.1 - 7 - 8 1053 75
AL833161.1 - 11 - 12 4121 75
BC020040.1 - 9 - 10 2295 75
BC037495.2 - 15 - 16 4650 75
BC038116.2 - 10 - 11 4313 75
BX648931.1 - 10 - 11 6267 75
D79994.2 - 11 - 12 4706 75

View Full Product Details