Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO38
Entrez Gene ID
81545
Gene Name
F-box protein 38
Gene Aliases
Fbx38, HMN2D, MOKA, SP329
Chromosome Location
Chr.5: 148383935 - 148442836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BI459318.1 - 3 - 4 337 125

View Full Product Details