Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX41
Entrez Gene ID
51428
Gene Name
DEAD-box helicase 41
Gene Aliases
ABS, MPLPF
Chromosome Location
Chr.5: 177511577 - 177517326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321732.1 NP_001308661.1 3 - 4 1026 66
NM_001321830.1 NP_001308759.1 4 - 5 818 66
NM_016222.3 NP_057306.2 4 - 5 749 66
GenBank mRNA AF195417.1 - 4 - 5 366 66
AK001255.1 - 4 - 5 374 66
AK027768.1 - 4 - 5 384 66
AK091545.1 - 3 - 4 1169 66
AK091774.1 - 3 - 4 661 66
AK222598.1 - 4 - 5 374 66
AK315491.1 - 4 - 5 384 66
BC015476.1 - 4 - 5 389 66
BX641072.1 - 2 - 3 433 66
DB039560.1 - 4 - 5 445 66

View Full Product Details