The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030630.2 | NP_085133.1 | 8 - 9 | 1158 | 77 |
XM_005257226.1 | XP_005257283.1 | 8 - 9 | 1153 | 77 | |
GenBank mRNA | AK074401.1 | - | 8 - 9 | 1087 | 77 |
AK304081.1 | - | 8 - 9 | 1042 | 77 | |
AL137556.1 | - | 8 - 9 | 1102 | 77 | |
BC032219.1 | - | 4 - 5 | 456 | 77 | |
BC035372.1 | - | 8 - 9 | 1074 | 77 |