The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003162.3 | NP_003153.2 | 13 - 14 | 1678 | 72 |
XM_005264519.4 | XP_005264576.1 | 12 - 13 | 1702 | 72 | |
XM_011533073.2 | XP_011531375.1 | 14 - 15 | 1900 | 72 | |
GenBank mRNA | AJ223814.1 | - | 13 - 14 | 1678 | 72 |
AK026934.1 | - | 10 - 11 | 1249 | 72 | |
BC106878.2 | - | 14 - 15 | 1595 | 72 | |
BC106879.2 | - | 13 - 14 | 1530 | 72 |