The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003162.3 | NP_003153.2 | 17 - 18 | 2181 | 147 |
XM_005264519.4 | XP_005264576.1 | 16 - 17 | 2205 | 147 | |
XM_011533073.2 | XP_011531375.1 | 18 - 19 | 2403 | 147 | |
GenBank mRNA | AJ223814.1 | - | 17 - 18 | 2181 | 147 |
AK026934.1 | - | 14 - 15 | 1752 | 147 | |
BC106878.2 | - | 18 - 19 | 2098 | 147 | |
BC106879.2 | - | 17 - 18 | 2033 | 147 |