The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003162.3 | NP_003153.2 | 5 - 6 | 670 | 66 |
XM_005264519.4 | XP_005264576.1 | 5 - 6 | 805 | 66 | |
XM_011533073.2 | XP_011531375.1 | 5 - 6 | 805 | 66 | |
GenBank mRNA | AJ223814.1 | - | 5 - 6 | 670 | 66 |
AK026934.1 | - | 2 - 3 | 241 | 66 | |
AK307836.1 | - | 1 - 2 | 347 | 66 | |
BC106878.2 | - | 6 - 7 | 587 | 66 | |
BC106879.2 | - | 6 - 7 | 633 | 66 |