Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNK1
Entrez Gene ID
8711
Gene Name
tyrosine kinase non receptor 1
Gene Aliases
-
Chromosome Location
Chr.17: 7380559 - 7389774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251902.1 NP_001238831.1 12 - 13 2126 107
NM_003985.4 NP_003976.2 12 - 13 2111 107
XM_011524045.2 XP_011522347.1 12 - 13 2159 107
XM_011524046.2 XP_011522348.1 12 - 13 2242 107
XM_011524047.2 XP_011522349.1 11 - 12 1949 107
XM_011524048.1 XP_011522350.1 12 - 13 2016 107
GenBank mRNA AK225403.1 - 12 - 13 1994 107
AK310684.1 - 13 - 14 2092 107
BC035782.1 - 11 - 12 1976 107
U43408.1 - 12 - 13 107

View Full Product Details