Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MTUS1
- Entrez Gene ID
-
57509
- Gene Name
- microtubule associated tumor suppressor 1
- Gene Aliases
-
ATBP, ATIP, ICIS, MP44, MTSG1
- Chromosome Location
-
Chr.8: 17643794 - 17801579 on Build GRCh38
View Full Product Details