The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005258145.1 | XP_005258202.2 | 16 - 17 | 2138 | 134 |
GenBank mRNA | AB037735.1 | - | 16 - 17 | 5089 | 134 |
AK001174.1 | - | 10 - 11 | 1145 | 134 | |
BC065274.1 | - | 14 - 15 | 1577 | 134 |