The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256440.1 | NP_001243369.1 | 7 - 8 | 721 | 73 |
NM_001256441.1 | NP_001243370.1 | 7 - 8 | 716 | 73 | |
NR_046202.1 | 7 - 8 | 769 | 73 | ||
XM_005258520.2 | XP_005258577.1 | 7 - 8 | 710 | 73 | |
XM_011526460.2 | XP_011524762.1 | 7 - 8 | 709 | 73 | |
XM_017026291.1 | XP_016881780.1 | 7 - 8 | 721 | 73 | |
XM_017026292.1 | XP_016881781.1 | 6 - 7 | 630 | 73 | |
XM_017026293.1 | XP_016881782.1 | 7 - 8 | 721 | 73 | |
XM_017026294.1 | XP_016881783.1 | 4 - 5 | 439 | 73 | |
XM_017026295.1 | XP_016881784.1 | 8 - 9 | 819 | 73 | |
GenBank mRNA | AK094207.1 | - | 7 - 8 | 709 | 73 |
AL834131.1 | - | 7 - 8 | 746 | 73 | |
BC023623.2 | - | 7 - 8 | 747 | 73 | |
BC027935.1 | - | 7 - 8 | 737 | 73 | |
BM459040.1 | - | 7 - 8 | 666 | 73 | |
CR936760.1 | - | 7 - 8 | 722 | 73 |