The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015541.2 | NP_056356.2 | 8 - 9 | 1088 | 64 |
XM_011533578.1 | XP_011531880.1 | 7 - 8 | 1545 | 64 | |
XM_011533579.2 | XP_011531881.1 | 8 - 9 | 888 | 64 | |
XM_017006134.1 | XP_016861623.1 | 7 - 8 | 1614 | 64 | |
XM_017006135.1 | XP_016861624.1 | 3 - 4 | 411 | 64 | |
XM_017006136.1 | XP_016861625.1 | 8 - 9 | 952 | 64 | |
GenBank mRNA | AB050468.1 | - | 8 - 9 | 1593 | 64 |
AF381545.1 | - | 8 - 9 | 1088 | 64 | |
AK314103.1 | - | 8 - 9 | 1599 | 64 | |
AY730707.1 | - | 8 - 9 | 1177 | 64 | |
BC071561.1 | - | 8 - 9 | 1678 | 64 | |
BC140907.1 | - | 8 - 9 | 1094 | 64 |