The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243142.1 | NP_001230071.1 | 9 - 10 | 1384 | 148 |
NM_001243143.1 | NP_001230072.1 | 9 - 10 | 1345 | 148 | |
NM_001301136.1 | NP_001288065.1 | 9 - 10 | 1387 | 148 | |
NM_016359.4 | NP_057443.2 | 9 - 10 | 1390 | 148 | |
NM_018454.7 | NP_060924.4 | 9 - 10 | 1387 | 148 | |
XM_005254428.2 | XP_005254485.1 | 9 - 10 | 1259 | 148 | |
XM_005254430.4 | XP_005254487.1 | 9 - 10 | 1181 | 148 | |
XM_006720559.2 | XP_006720622.1 | 9 - 10 | 1262 | 148 | |
XM_006720560.3 | XP_006720623.1 | 9 - 10 | 1358 | 148 | |
XM_006720561.2 | XP_006720624.1 | 9 - 10 | 1176 | 148 | |
XM_017022295.1 | XP_016877784.1 | 9 - 10 | 2054 | 148 | |
GenBank mRNA | AF090915.1 | - | 9 - 10 | 1193 | 148 |
AF290612.1 | - | 9 - 10 | 1194 | 148 | |
AF305711.1 | - | 9 - 10 | 1123 | 148 | |
AK023483.1 | - | 9 - 10 | 1197 | 148 | |
AK222819.1 | - | 9 - 10 | 1211 | 148 | |
AK290879.1 | - | 9 - 10 | 1214 | 148 | |
AK307728.1 | - | 9 - 10 | 1184 | 148 | |
AL833611.1 | - | 9 - 10 | 1157 | 148 | |
BC001308.1 | - | 9 - 10 | 1190 | 148 | |
BC010838.1 | - | 7 - 8 | 931 | 148 | |
BC012887.1 | - | 9 - 10 | 1168 | 148 | |
BC024772.1 | - | 9 - 10 | 1192 | 148 |