The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006772.2 | NP_006763.2 | 14 - 15 | 2525 | 64 |
GenBank mRNA | AL713634.1 | - | 7 - 8 | 1181 | 64 |