Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
Chromosome Location
Chr.19: 18001132 - 18014102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286826.1 NP_001273755.1 1 - 2 418 121
NM_015683.1 NP_056498.1 1 - 2 418 121
GenBank mRNA AF193051.1 - 1 - 2 418 121
AK314657.1 - 1 - 2 416 121
BC022516.1 - 1 - 2 416 121

View Full Product Details