Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
Chromosome Location
Chr.19: 18001132 - 18014102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025604.2 NP_001020775.1 3 - 4 661 74
NM_001286826.1 NP_001273755.1 3 - 4 636 74
NM_015683.1 NP_056498.1 3 - 4 636 74
GenBank mRNA AF131826.1 - 2 - 3 168 74
AF193051.1 - 3 - 4 636 74
AK000689.1 - 3 - 4 634 74
AK127940.1 - 3 - 4 625 74
AK314657.1 - 3 - 4 634 74
BC022516.1 - 3 - 4 634 74

View Full Product Details