The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058243.2 | NP_490597.1 | 18 - 19 | 4004 | 75 |
XM_011527854.1 | XP_011526156.1 | 18 - 19 | 4069 | 75 | |
GenBank mRNA | AF386649.1 | - | 18 - 19 | 4004 | 75 |
BC008354.1 | - | 2 - 3 | 236 | 75 |