The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303040.1 | NP_001289969.1 | 10 - 11 | 1448 | 88 |
NM_001303041.1 | NP_001289970.1 | 9 - 10 | 1336 | 88 | |
NM_003637.4 | NP_003628.2 | 13 - 14 | 1765 | 88 | |
XM_005277436.3 | XP_005277493.1 | 12 - 13 | 1516 | 88 | |
XM_011510083.2 | XP_011508385.1 | 9 - 10 | 1196 | 88 | |
XM_011510084.1 | XP_011508386.1 | 2 - 3 | 193 | 88 | |
XM_017002622.1 | XP_016858111.1 | 11 - 12 | 1722 | 88 | |
XM_017002624.1 | XP_016858113.1 | 11 - 12 | 1722 | 88 | |
XM_017002625.1 | XP_016858114.1 | 11 - 12 | 1722 | 88 | |
XM_017002626.1 | XP_016858115.1 | 9 - 10 | 1479 | 88 | |
XM_017002627.1 | XP_016858116.1 | 11 - 12 | 1722 | 88 | |
XM_017002628.1 | XP_016858117.1 | 7 - 8 | 1172 | 88 | |
GenBank mRNA | AF074015.1 | - | 13 - 14 | 1611 | 88 |
AF112345.1 | - | 13 - 14 | 1666 | 88 | |
AK304155.1 | - | 10 - 11 | 1304 | 88 | |
AK304183.1 | - | 12 - 13 | 1595 | 88 | |
AK314255.1 | - | 13 - 14 | 1614 | 88 | |
BC140831.1 | - | 9 - 10 | 1211 | 88 | |
BC144637.1 | - | 9 - 10 | 1211 | 88 |