The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079843.2 | NP_001073312.1 | 9 - 10 | 2009 | 60 |
NM_017766.4 | NP_060236.3 | 9 - 10 | 2009 | 60 | |
XM_005263479.3 | XP_005263536.1 | 7 - 8 | 1873 | 60 | |
XM_011541635.2 | XP_011539937.1 | 7 - 8 | 1872 | 60 | |
XM_017001539.1 | XP_016857028.1 | 7 - 8 | 1873 | 60 | |
XM_017001540.1 | XP_016857029.1 | 9 - 10 | 1983 | 60 | |
XM_017001541.1 | XP_016857030.1 | 9 - 10 | 2045 | 60 | |
GenBank mRNA | AK000328.1 | - | 5 - 6 | 1323 | 60 |
AK092289.1 | - | 9 - 10 | 1983 | 60 | |
AK225324.1 | - | 5 - 6 | 1323 | 60 | |
AK293839.1 | - | 7 - 8 | 1841 | 60 | |
BC051883.2 | - | 6 - 7 | 1679 | 60 | |
DQ217660.1 | - | 9 - 10 | 1981 | 60 |